APO455837215 Apollo Scientific Ltd

2-Amino-4-fluorobenzonitrile

  • MDL number MFCD09955451
  • Cas Number : 80517-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Synonym 4-Fluoroanthranilonitrile, 2-Cyano-5-fluoroaniline
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-fluorobenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-fluorobenzonitrile
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart