APO455837216 Apollo Scientific Ltd

4-Fluoro-2-iodobenzonitrile

  • MDL number MFCD09998181
  • Cas Number : 1031929-20-0
  • Molecular Weight : 247.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-2-iodobenzonitrile

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-2-iodobenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart