APO455837218 Apollo Scientific Ltd

5-Bromo-2,2-difluoro-1,3-benzodioxole

  • MDL number MFCD00236212
  • Cas Number : 33070-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1,2-(difluoromethylenedioxy)benzene
Purity 98
Density 1.74
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 78-79/20mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=486

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=486
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart