APO455837222 Apollo Scientific Ltd

2-(N-Methylhydrazino)-5-(trifluoromethyl)pyridine

  • MDL number MFCD00052297
  • Cas Number : 163620-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-55
Synonym N-Methyl-N-[5-(trifluoromethyl)pyridin-2-yl]hydrazine, 6-(N-Methylhydrazino)-alpha,alpha,alpha-trifluoro-3-picoline
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5479

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5479
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart