APO455837226 Apollo Scientific Ltd

2,4,6-Trifluoro-1-bromo-3,5-dichlorobenzene

  • MDL number MFCD03424469
  • Cas Number : 24812-13-3
  • Molecular Weight : 279.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-59.5
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/246-trifluoro-1-bromo-35-dichlorobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/246-trifluoro-1-bromo-35-dichlorobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart