APO455837230 Apollo Scientific Ltd

Methyl 3,3,3-trifluoropropanoate

  • MDL number MFCD02262206
  • Cas Number : 18830-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3,3,3-trifluoropropionate
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 95-96
Product Link https://store.apolloscientific.co.uk/product/methyl-333-trifluoropropanoate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-333-trifluoropropanoate
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart