APO455837239 Apollo Scientific Ltd

2-Fluoro-6-nitrobenzoic acid

  • MDL number MFCD01862079
  • Cas Number : 385-02-4
  • Molecular Weight : 185.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-147
Synonym 2-Carboxy-3-fluoronitrobenzene
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-nitrobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart