APO455837246 Apollo Scientific Ltd

2,3,4,5,6-Pentafluorobiphenyl

  • MDL number MFCD00093108
  • Cas Number : 784-14-5
  • Molecular Weight : 244.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-113
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23456-pentafluorobiphenyl

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23456-pentafluorobiphenyl
pictograms GHS07
hstatements H315;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart