APO455837249 Apollo Scientific Ltd

1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxamide

  • MDL number MFCD03237056
  • Cas Number : 465514-29-8
  • Molecular Weight : 274.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-trifluoromethylpyrimidin-2-ylpiperidine-4-carboxamide

Safety Information

signalword Warning
pcodes P280;P261
MSDS https://store.apolloscientific.co.uk/product/1-4-trifluoromethylpyrimidin-2-ylpiperidine-4-carboxamide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart