APO455837250 Apollo Scientific Ltd

Bromo(difluoro)acetic acid

  • MDL number MFCD02183519
  • Cas Number : 354-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 26.5
Synonym 2-Bromo-2,2-difluoroethanoic acid
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 134
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2423

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=2423
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart