APO455837253 Apollo Scientific Ltd

3-Amino-2-chloro-6-(trifluoromethyl)pyridine

  • MDL number MFCD01862661
  • Cas Number : 117519-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-94
Synonym 2-Chloro-6-(trifluoromethyl)pyridin-3-amine, 5-Amino-6-chloro-alpha,alpha,alpha-trifluoro-2-picoline
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-2-chloro-6-trifluoromethylpyridine-98

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-2-chloro-6-trifluoromethylpyridine-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart