APO455837259 Apollo Scientific Ltd

1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carbonyl chloride

  • MDL number MFCD00173866
  • Cas Number : 175137-36-7
  • Molecular Weight : 319.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-79
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-nitrophenyl-5-trifluoromethylpyrazole-4-carbonyl-chloride

Safety Information

signalword Danger
pcodes P280;P301+P330+P331;P304+P340
MSDS https://store.apolloscientific.co.uk/product/1-4-nitrophenyl-5-trifluoromethylpyrazole-4-carbonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart