APO455837264 Apollo Scientific Ltd

1-[5-(Trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid

  • MDL number MFCD00209248
  • Cas Number : 406476-31-1
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 162-164
Iupac Name 1-[5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
Inchi Code 1S/C12H13F3N2O2/c13-12(14,15)9-1-2-10(16-7-9)17-5-3-8(4-6-17)11(18)19/h1-2,7-8H,3-6H2,(H,18,19)
Inchi Key KNDSIDUPVUCATQ-UHFFFAOYSA-N

Description

Synonym 2-(4-Carboxypiperidin-1-yl)-5-(trifluoromethyl)pyridine, 4-Carboxy-1-[5-(trifluoromethyl)pyridin-2-yl]piperidine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 162-164
Product Link https://store.apolloscientific.co.uk/product/1-5-trifluoromethylpyridin-2-ylpiperidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-5-trifluoromethylpyridin-2-ylpiperidine-4-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart