APO455837277 Apollo Scientific Ltd

4-Methoxymetanilyl fluoride

  • MDL number MFCD00007417
  • Cas Number : 498-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-63
Synonym 3-Amino-4-methoxybenzenesulphonyl fluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3837

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3837
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart