APO455837292 Apollo Scientific Ltd

1-(Pentafluorophenyl)ethanol

  • MDL number MFCD00004510
  • Cas Number : 830-50-2
  • Molecular Weight : 212.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32-34
Synonym alpha-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol, Methyl pentafluorophenyl carbinol
Purity 97
Flash Point 87
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68-70/30mm
Product Link https://store.apolloscientific.co.uk/product/1-pentafluorophenylethanol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-pentafluorophenylethanol
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart