APO455837304 Apollo Scientific Ltd

5-(Trifluoromethoxy)-1H-indazole, N1-BOC protected

  • MDL number MFCD20481820
  • Cas Number : 1346521-27-4
  • Molecular Weight : 302.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 5-(trifluoromethoxy)-1H-indazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-5-(trifluoromethoxy)-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-trifluoromethoxy-1h-indazole-n1-boc-protected

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-trifluoromethoxy-1h-indazole-n1-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart