APO455837307 Apollo Scientific Ltd

3,5-Difluoro-4-nitrobromobenzene

  • MDL number MFCD12025434
  • Cas Number : 147808-42-2
  • Molecular Weight : 237.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-1,3-difluoro-2-nitrobenzene
Inchi Code 1S/C6H2BrF2NO2/c7-3-1-4(8)6(10(11)12)5(9)2-3/h1-2H
Inchi Key ABAGKHWCTWMUDH-UHFFFAOYSA-N

Description

Synonym 5-Bromo-1,3-difluoro-2-nitrobenzene, 4-Bromo-2,6-difluoronitrobenzene
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-4-nitrobromobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-4-nitrobromobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart