APO455837308 Apollo Scientific Ltd

Methyl 2-chloro-5-fluoroisonicotinate

  • MDL number MFCD09909836
  • Cas Number : 876919-10-7
  • Molecular Weight : 189.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 2-chloro-5-fluoropyridine-4-carboxylate, 2-Chloro-5-fluoro-4-(methoxycarbonyl)pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-chloro-5-fluoroisonicotinate

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-chloro-5-fluoroisonicotinate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart