APO455837313 Apollo Scientific Ltd

2,6-Difluoro-3-nitroaniline

  • MDL number MFCD11110684
  • Cas Number : 25892-09-5
  • Molecular Weight : 174.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Amino-2,4-difluoronitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-nitroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-nitroaniline
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart