APO455837317 Apollo Scientific Ltd

2-Fluoro-4-nitrobenzyl alcohol

  • MDL number MFCD11110168
  • Cas Number : 660432-43-9
  • Molecular Weight : 171.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Synonym 3-Fluoro-4-(hydroxymethyl)nitrobenzene, (2-Fluoro-4-nitrophenyl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-nitrobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart