APO455837321 Apollo Scientific Ltd

3-Fluoro-4-iodobenzyl alcohol

  • MDL number MFCD18396947
  • Cas Number : 1261615-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Fluoro-4-iodophenyl)methanol
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=62641

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=62641
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart