APO455837322 Apollo Scientific Ltd

6-Bromo-3-chloro-2-fluorobenzaldehyde

  • MDL number MFCD11520661
  • Cas Number : 1114809-02-7
  • Molecular Weight : 237.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-3-chloro-2-fluorobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-3-chloro-2-fluorobenzaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart