APO455837328 Apollo Scientific Ltd

4-Bromo-3-fluorobenzyl bromide

  • MDL number MFCD11847179
  • Cas Number : 127425-73-4
  • Molecular Weight : 267.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-44
Synonym 1-Bromo-4-(bromomethyl)-2-fluorobenzene, alpha,4-Dibromo-3-fluorotoluene
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 112-114/8mm
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-fluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart