APO455837330 Apollo Scientific Ltd

Methyl 3-fluorobenzoate

  • MDL number MFCD03094302
  • Cas Number : 455-68-5
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -10
Purity 97
Density 1.20
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 197
Product Link https://store.apolloscientific.co.uk/product/methyl-3-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart