APO455837331 Apollo Scientific Ltd

Perfluoropropanamide

  • MDL number MFCD00039771
  • Cas Number : 354-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95
Synonym 2,2,3,3,3-Pentafluoropropanamide, 2,2,3,3,3-Pentafluoropropionamide,
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4322

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4322
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart