APO455837332 Apollo Scientific Ltd

2-Hydroxy-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD07787570
  • Cas Number : 79427-88-6
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-4-(trifluoromethyl)phenol, 3-Carboxy-4-hydroxybenzotrifluoride, 5-(Trifluoromethyl)salicylic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-5-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-5-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart