APO455837333 Apollo Scientific Ltd

4-Bromo-3-(trifluoromethyl)benzaldehyde

  • MDL number MFCD08059130
  • Cas Number : 34328-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-54
Synonym 2-Bromo-5-formylbenzotrifluoride, 4-Bromo-alpha,alpha,alpha-trifluoro-m-tolualdehyde
Purity 99
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45649

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45649
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart