APO455837338 Apollo Scientific Ltd

2-(Trifluoromethyl)pyridine-4-boronic acid

  • MDL number MFCD11616522
  • Cas Number : 1093407-58-9
  • Molecular Weight : 190.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Borono-2-(trifluoromethyl)pyridine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-trifluoromethylpyridine-4-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoromethylpyridine-4-boronic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart