APO455837355 Apollo Scientific Ltd

2-(2H-Tetrafluoroethoxy)aniline

  • MDL number MFCD03094306
  • Cas Number : 35295-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(1,1,2,2-Tetrafluoroethoxy)aniline
Purity 98
Density 1.7
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 92/12mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5900

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5900
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart