APO455837364 Apollo Scientific Ltd

trans-3-Fluoro-5-(trifluoromethyl)cinnamic acid

  • MDL number MFCD00236296
  • Cas Number : 575469-96-4
  • Molecular Weight : 234.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2E)-3-[3-Fluoro-5-(trifluoromethyl)phenyl]prop-2-enoic acid, (E)-3-[3-Fluoro-5-(trifluoromethyl)phenyl]acrylic acid
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/trans-3-fluoro-5-trifluoromethylcinnamic-acid

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-3-fluoro-5-trifluoromethylcinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart