APO455837365 Apollo Scientific Ltd

5-Bromo-2-(difluoromethoxy)toluene

  • MDL number MFCD11036069
  • Cas Number : 888327-32-0
  • Molecular Weight : 237.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1-(difluoromethoxy)-2-methylbenzene
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-difluoromethoxytoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-difluoromethoxytoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart