APO455837372 Apollo Scientific Ltd

1-[2-(Trifluoromethyl)quinol-4-yl]piperazine

  • MDL number MFCD00219765
  • Cas Number : 175203-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-135
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-2-trifluoromethylquinol-4-ylpiperazine-97

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-trifluoromethylquinol-4-ylpiperazine-97
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart