APO455837377 Apollo Scientific Ltd

3,3,4,4,5,5,6,6,6-Nonafluorohex-1yl acrylate

  • MDL number MFCD00236104
  • Cas Number : 52591-27-2
  • Molecular Weight : 318.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -68
Synonym 2-(Perfluorobut-1-yl)ethyl acrylate, 3,3,4,4,5,5,6,6,6-Nonafluorohex-1-yl prop-2-enoate, TEAc-4
Purity 97
Density 1.44
Flash Point 52
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 160
Product Link https://store.apolloscientific.co.uk/product/334455666-nonafluorohex-1yl-acrylate

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/334455666-nonafluorohex-1yl-acrylate
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart