APO455837378 Apollo Scientific Ltd

1H,1H,2H,2H-Perfluorohexyl methacrylate

  • MDL number MFCD00236094
  • Cas Number : 1799-84-4
  • Molecular Weight : 332.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Nonafluorobutyl)ethyl methacrylate
Purity 97
Density 1.40
Flash Point 77
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60-62/5mm
Product Link https://store.apolloscientific.co.uk/product/1h1h2h2h-perfluorohexyl-methacrylate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/1h1h2h2h-perfluorohexyl-methacrylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart