APO455837381 Apollo Scientific Ltd

2-Fluoro-4-methylbenzonitrile

  • MDL number MFCD03094311
  • Cas Number : 85070-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Synonym 4-Cyano-3-fluorotoluene, 2-Fluoro-p-tolunitrile
Purity 98
Flash Point 80
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methylbenzonitrile-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methylbenzonitrile-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart