APO455837382 Apollo Scientific Ltd

3,5-Difluorotoluene

  • MDL number MFCD00797323
  • Cas Number : 117358-51-7
  • Molecular Weight : 128.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-difluoro-5-methylbenzene
Inchi Code 1S/C7H6F2/c1-5-2-6(8)4-7(9)3-5/h2-4H,1H3
Inchi Key YISYUYYETHYYMD-UHFFFAOYSA-N

Description

Synonym 1,3-Difluoro-5-methylbenzene
Purity 98%
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluorotoluene

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluorotoluene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart