APO455837383 Apollo Scientific Ltd

3-Fluoro-5-methylbenzonitrile

  • MDL number MFCD03094312
  • Cas Number : 216976-30-6
  • Molecular Weight : 135.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Synonym 3-Cyano-5-fluorotoluene, 5-Fluoro-m-tolunitrile
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methylbenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart