APO455837384 Apollo Scientific Ltd

3-Fluoro-5-methylbenzoic acid

  • MDL number MFCD03094313
  • Cas Number : 518070-19-4
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-145
Synonym 3-Carboxy-5-fluorotoluene, 5-Fluoro-m-toluic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart