APO455837386 Apollo Scientific Ltd

3-Fluoro-5-methylbenzaldehyde

  • MDL number MFCD03094315
  • Cas Number : 189628-39-5
  • Molecular Weight : 138.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-5-methylbenzaldehyde
Inchi Code 1S/C8H7FO/c1-6-2-7(5-10)4-8(9)3-6/h2-5H,1H3
Inchi Key XXPITUUQKCGWNR-UHFFFAOYSA-N

Description

Synonym 5-Fluoro-m-tolualdehyde, 3-Fluoro-5-formyltoluene
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-5-methylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-5-methylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart