APO455837391 Apollo Scientific Ltd

5-Cyano-2-fluorobenzoic acid

  • MDL number MFCD03094321
  • Cas Number : 146328-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-189
Synonym 3-Carboxy-4-fluorobenzonitrile
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5916

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5916
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart