APO455837393 Apollo Scientific Ltd

4-(Bromomethyl)-2-fluorobenzonitrile

  • MDL number MFCD03094323
  • Cas Number : 222978-03-2
  • Molecular Weight : 214.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-46
Synonym 4-Cyano-3-fluorobenzyl bromide, alpha-Bromo-2-fluoro-p-tolunitrile
Purity 98
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromomethyl-2-fluorobenzonitrile

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromomethyl-2-fluorobenzonitrile
pictograms GHS05;GHS07;GHS08
hstatements H314;H334
Availability details for
Quantity
  • Availability
  • Add to Cart