APO455837395 Apollo Scientific Ltd

Perfluorocyclohexanecarbonyl fluoride

  • MDL number MFCD00236697
  • Cas Number : 6588-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Undecafluorocyclohexanecarbonyl fluoride, (Fluorocarbonyl)perfluorocyclohexane
Purity 65
Density 1.6
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 70
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4352

Safety Information

signalword Danger
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4352
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart