APO455837396 Apollo Scientific Ltd

2-Fluoro-4-methylbenzaldehyde

  • MDL number MFCD03094325
  • Cas Number : 146137-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluoro-4-formyltoluene, 2-Fluoro-p-tolualdehyde
Purity 98
Density 0.95
Flash Point 84
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 91-92/14mm
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methylbenzaldehyde-98

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methylbenzaldehyde-98
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart