APO455837397 Apollo Scientific Ltd

2-Fluoro-4-methylbenzoic acid

  • MDL number MFCD03092906
  • Cas Number : 7697-23-6
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-189
Synonym 4-Carboxy-3-fluorotoluene, 2-Fluoro-p-toluic acid
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart