APO455837415 Apollo Scientific Ltd

1H-Perfluorodecane

  • MDL number MFCD00042110
  • Cas Number : 375-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31-33
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 159-160
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4361

Safety Information

signalword Warning
pcodes P302+P352;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4361
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart