APO455837426 Apollo Scientific Ltd

2,5-Bis(trifluoromethyl)phenylacetonitrile

  • MDL number MFCD00799489
  • Cas Number : 302911-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,5-Bis(trifluoromethyl)benzyl cyanide
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2389

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2389
pictograms GHS06
hstatements H311;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart