APO455837432 Apollo Scientific Ltd

2-Chloro-4-fluoropyridine

  • MDL number MFCD04039345
  • Cas Number : 34941-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.456
Flash Point 54
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 140
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluoropyridine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluoropyridine
pictograms GHS02;GHS07
hstatements H226;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart