APO455837435 Apollo Scientific Ltd

1H,1H,2H,2H-Perfluorododecan-1-ol

  • MDL number MFCD00039545
  • Cas Number : 865-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-95
Synonym 2-(Perfluorodecyl)ethanol, 1H,1H,2H,2H-Henicosafluorododecan-1-ol
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 145/10mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4396

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4396
pictograms GHS02;GHS07
hstatements H228;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart