APO455837441 Apollo Scientific Ltd

2,3,5-Trifluorobenzonitrile

  • MDL number MFCD00083539
  • Cas Number : 241154-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.36
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5028

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5028
pictograms GHS06
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart