APO455837444 Apollo Scientific Ltd

4-Bromo-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD06659466
  • Cas Number : 191165-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-cyanobenzotrifluoride, 4-Bromo-alpha,alpha,alpha-trifluoro-o-tolunitrile
Purity 98
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45907

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=45907
Availability details for
Quantity
  • Availability
  • Add to Cart